Anti-Golgin subfamily A member 4/Golgin 245 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-13487R-A750
BOSSBS-13487R-A750EA 890 EUR
BOSSBS-13487R-A750
Anti-Golgin subfamily A member 4/Golgin 245 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Golgin 245 is also known as p230, GCP2, GOLG or golgi autoantigen and is a 2,230 amino acid protein that is expressed as three isoforms. Golgin 245 is localized to the cytoplasm in cells and is a member of the golgin family, all of which are peripheral membrane proteins associated with the Golgi complex. Golgin 245 has a carboxyl-terminal GRIP domain, which attaches to the trans-Golgi network (TGN) and TGN-derived vesicles. It is thought that the interaction that takes place between golgin 245 and MACF1, which cross-links microtubules to the Actin cytoskeleton, allows proteins to be transported from the TGN to the cell periphery. against golgin 245 are associated with two chronic disorders known as Sjren's syndrome and Hepatitis B.
Type: Primary
Antigen: Golgin subfamily A member 4/Golgin 245
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 2061-2160/2230
Host: Rabbit
Isotype: IgG
Reactivity: Mouse