Anti-GPAT2 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-13497R-CY5
BOSSBS-13497R-CY5EA 900 EUR
BOSSBS-13497R-CY5
Anti-GPAT2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Esterifies acyl-group from acyl-ACP to the sn-1 position of glycerol-3-phosphate, an essential step in glycerolipid biosynthesis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GPAT2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat