Anti-GPR27 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-13528R
BOSSBS-13528REA 725 EUR
BOSSBS-13528R
Anti-GPR27 Rabbit Polyclonal Antibody
Antikörper
G protein-coupled receptors (GPRs) are a protein family of transmembrane receptors that transmit an extracellular signal (ligand binding) into an intracellular signal (G protein activation). GPR signaling is an evolutionarily ancient mechanism used by all eukaryotes to sense environmental stimuli and mediate cell-cell communication. GPRs all have seven membrane-spanning domains and extracellular loops that can be glycosylated. These extracellular loops also contain two highly conserved cysteine residues which create disulfide bonds to stabilize the receptor structure. SREB1 (super conserved receptor expressed in brain 1), also known as GPR27 (G protein-coupled receptor 27), belongs to the SREB subfamily of GPRs that are expressed in the central nervous system. SREB1 may function as an amine-like GPR.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: GPR27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR