Anti-PTGDR2 Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-13537R-CY7
BOSSBS-13537R-CY7EA 900 EUR
BOSSBS-13537R-CY7
Anti-PTGDR2 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Receptor for prostaglandin D2 (PGD2). Coupled to the G(i)-protein. Receptor activation may result in pertussis toxin-sensitive decreases in cAMP levels and Ca(2+) mobilization. PI3K signaling is also implicated in mediating PTGDR2 effects. PGD2 induced receptor internalization. CRTH2 internalization can be regulated by diverse kinases such as, PKC, PKA, ADRBK1/GRK2, GPRK5/GRK5 and GRK6. Receptor activation is responsible, at least in part, in immune regulation and allergic/inflammation responses.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PTGDR2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat