Anti-ZBTB22 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-13566R-CY3
BOSSBS-13566R-CY3EA 900 EUR
BOSSBS-13566R-CY3
Anti-ZBTB22 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
ZBTB22 contains 1 BTB (POZ) domain and 3 C2H2-type zinc fingers and belongs to the krueppel C2H2-type zinc finger protein family. ZBTB22 may be involved in transcriptional regulation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZBTB22
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR