Anti-ZBTB39 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-13573R
BOSSBS-13573REA 725 EUR
BOSSBS-13573R
Anti-ZBTB39 Rabbit Polyclonal Antibody
Antikörper
ZBTB39 may be involved in transcriptional regulation. It belongs to the krueppel C2H2-type zinc-finger protein family and contains one BTB (POZ) domain and eight C2H2-type zinc fingers.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: ZBTB39
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR