Anti-ZBTB39 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-13573R-A680
BOSSBS-13573R-A680EA 890 EUR
BOSSBS-13573R-A680
Anti-ZBTB39 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
ZBTB39 may be involved in transcriptional regulation. It belongs to the krueppel C2H2-type zinc-finger protein family and contains one BTB (POZ) domain and eight C2H2-type zinc fingers.

Type: Primary
Antigen: ZBTB39
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1-100/712
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR