Anti-ZBTB39 Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-13573R-CY7
BOSSBS-13573R-CY7EA 900 EUR
BOSSBS-13573R-CY7
Anti-ZBTB39 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
ZBTB39 may be involved in transcriptional regulation. It belongs to the krueppel C2H2-type zinc-finger protein family and contains one BTB (POZ) domain and eight C2H2-type zinc fingers.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ZBTB39
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat