Anti-ZBTB48/ZNF855 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-13580R-A750
BOSSBS-13580R-A750EA 890 EUR
BOSSBS-13580R-A750
Anti-ZBTB48/ZNF855 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kr_ppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. Zinc finger and BTB domain-containing protein 48 (ZBTB48), also known as Kruppel-related zinc finger protein 3 or HKR3, is a 688 amino acid member of the Kr_ppel C2H2-type zinc-finger protein family. Localized to the nucleus, ZBTB48 is expressed in the adrenal gland and neuroblastoma cell lines. ZBTB48 contains a BTB domain, also known as a POZ domain, which inhibits DNA binding and mediates homotypic and heterotypic dimerization. Characteristics of the BTB domain suggest that ZBTB48 functions as a transcription regulator.

Type: Primary
Antigen: ZBTB48/ZNF855
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 451-550/688
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR