Anti-ZNF577 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-13588R-A647
BOSSBS-13588R-A647EA 893 EUR
BOSSBS-13588R-A647
Anti-ZNF577 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Krüppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. Zinc finger protein 575 (ZNF575) is a 245 amino acid member of the Krüppel C2H2-type zinc-finger protein family. Localized to the nucleus, ZNF575 contains six C2H2-type zinc fingers through which it is thought to be involved in DNA-binding and transcriptional regulation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZNF577
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR