Anti-SLAIN1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-13602R-HRP
BOSSBS-13602R-HRPEA 900 EUR
BOSSBS-13602R-HRP
Anti-SLAIN1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
SLAIN1 is a 561 amino acid member of the SLAIN motif-containing family and is expressed in embryonic stem cells, as well as in lung, brain and testis, where it exists as two alternatively spliced isoforms. The gene encoding SLAIN1 maps to human chromosome 13, which houses over 400 genes, such as BRCA2 and RB1, and comprises nearly 4% of the human genome. As with most chromosomes, polysomy of part or all of chromosome 13 is deleterious to development and decreases the odds of survival. Trisomy 13, also known as Patau syndrome, is deadly and the few who survive past one year suffer from permanent neurologic defects, difficulty eating and vulnerability to serious respiratory infections.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: SLAIN1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR