Anti-TMEM176A Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-13623R-A680
BOSSBS-13623R-A680EA 890 EUR
BOSSBS-13623R-A680
Anti-TMEM176A Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
In mice, this protein is preferentially expressed in tissues of lymphoid nature, such as lymph node, thymus, bone marrow and spleen. This protein is localized to intracellular compartments and plasma membrane.

Type: Primary
Antigen: TMEM176A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 21-120/245
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR