Anti-JIP2 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-13647R-CY3
BOSSBS-13647R-CY3EA 900 EUR
BOSSBS-13647R-CY3
Anti-JIP2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
c-Jun NH2-terminal kinases (JNKs) are distant members of the MAP kinase family (1). JNK1 is activated by dual phosphorylation at a Thr-Pro-Tyr motif in response to ultraviolet (UV) light, and it functions to phosphorylate c-Jun at amino terminal serine regulatory sites, Ser-63 and Ser-73, resulting in transcriptional activation (2-5). Two additional JNK family members have been identified as JNK2 and JNK3 (3). JIP-1 (for JNK interacting protein-1) has been identified as a cytoplasmic inhibitor of JNK that retains JNK in the cytoplasm, thereby inhibiting JNK-regulated gene expression. Evidence suggests that JNK1 and JNK2 bind to JIP-1 with greater affinity than to ATF-2 and c-Jun, which are targets of the JNK signaling pathway. JIP-1 contains an amino terminal JNK binding domain and a carboxy terminal SH3 domain. ATF-2 and c-Jun also contain the JNK binding domain and are thought to compete with JIP-1 for JNK binding (6). Multiple splice variants if JIP-1, including JIP-1b, JIP-1c (also designated islet-brain 1 or IB-1), JIP-2a, JIP-2b and JIP-3, have been identified in brain (7).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: JIP2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR