Anti-LIMS2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-13650R-A350
BOSSBS-13650R-A350EA 893 EUR
BOSSBS-13650R-A350
Anti-LIMS2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Pinch is a focal adhesion protein that is a component of the ILK-PINCH complex. This complex is a major part of the growth factor and integrin signaling pathway. Pinch is involved in cell differentiation, proliferation and survival by acting as an effector of integrin and growth factor signaling. It is a cytoplasmic protein expressed in most tissues and consists of five LIM domains, a nuclear localization signal and a nuclear export signal. The PINCH-1/ILK complex is regulated by another member of the Pinch family, PINCH-2, which also forms a complex with ILK.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LIMS2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR