Anti-LIMS2 Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-13650R-CY3
BOSSBS-13650R-CY3EA 900 EUR
BOSSBS-13650R-CY3
Anti-LIMS2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Pinch is a focal adhesion protein that is a component of the ILK-PINCH complex. This complex is a major part of the growth factor and integrin signaling pathway. Pinch is involved in cell differentiation, proliferation and survival by acting as an effector of integrin and growth factor signaling. It is a cytoplasmic protein expressed in most tissues and consists of five LIM domains, a nuclear localization signal and a nuclear export signal. The PINCH-1/ILK complex is regulated by another member of the Pinch family, PINCH-2, which also forms a complex with ILK.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LIMS2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse