Anti-TIFA Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
BS-13669R-A350
BOSSBS-13669R-A350EA 893 EUR
BOSSBS-13669R-A350
Anti-TIFA Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Adapter protein which mediates the IRAK1 and TRAF6 interaction following IL-1 stimulation, resulting in the downstream activation of NF-kappa-B and AP-1 pathways. Induces the oligomerization and polyubiquitination of TRAF6, which leads to the activation of TAK1 and IKK through a proteasome-independent mechanism.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TIFA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human