Anti-TIFA Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-13669R-A555
BOSSBS-13669R-A555EA 893 EUR
BOSSBS-13669R-A555
Anti-TIFA Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Adapter protein which mediates the IRAK1 and TRAF6 interaction following IL-1 stimulation, resulting in the downstream activation of NF-kappa-B and AP-1 pathways. Induces the oligomerization and polyubiquitination of TRAF6, which leads to the activation of TAK1 and IKK through a proteasome-independent mechanism.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TIFA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR