Anti-TMEM115 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-13697R
BOSSBS-13697REA 725 EUR
BOSSBS-13697R
Anti-TMEM115 Rabbit Polyclonal Antibody
Antikörper
Transmembrane proteins contain transmembrane domains that are usually characterized by alpha-helical structures. Transmembrane proteins exist as thermodynamically stable hetero- and homodimers that interact with the lipid bilayer and are involved in both material exchange and communication between the cell and the environment. PL6 is a 351 amino acid multi-pass membrane protein that is highly expressed in kidney and skeletal muscle with lower levels of expression detected in liver, placenta, prancreas, lung, heart and brain. PL6 contains one phosphoserine residue and several transmembraneMore Product Information
Bestellen

TECHNISCHE DATEN

Antigen-Symbol TMEM115
Antigen-Name Transmembrane Protein 115
Environmentally Preferable
Antigen-Synonyme PP6|Placental protein 6.
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Human,Mouse
Wirt Rabbit
Isotyp IgG
Western blot Yes
ImmunoChemie Yes
Form liquid
Reinigung Purified by Protein A.
Konzentration 1 ug/ul
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Transporttemperatur 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR