Anti-Claudin 20 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-13748R-HRP
BOSSBS-13748R-HRPEA 893 EUR
BOSSBS-13748R-HRP
Anti-Claudin 20 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Plays a major role in tight junction-specific obliteration of the intercellular space, through calcium-independent cell-adhesion activity.

Type: Primary
Antigen: Claudin 20
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 41-140/219
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR