Anti-Claudin 19 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-13749R
BOSSBS-13749REA 719 EUR
BOSSBS-13749R
Anti-Claudin 19 Rabbit Polyclonal Antibody
Antikörper
Plays a major role in tight junction-specific obliteration of the intercellular space, through calcium-independent cell-adhesion activity.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Claudin 19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR