Anti-Claudin 19 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-13749R-A488
BOSSBS-13749R-A488EA 893 EUR
BOSSBS-13749R-A488
Anti-Claudin 19 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Plays a major role in tight junction-specific obliteration of the intercellular space, through calcium-independent cell-adhesion activity.

Type: Primary
Antigen: Claudin 19
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 488
Public Immunogen Range: 21-120/224
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR