Anti-Ghrelin 28 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-1375R-A680
BOSSBS-1375R-A680EA 874 EUR
BOSSBS-1375R-A680
Anti-Ghrelin 28 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Ghrelin is an endogenous ligand for the growth hormone secretagogue receptor and is involved in regulating growth hormone release. Ghrelin is derived from a preprohormone called preproghrelin, which also generates a second peptide called obestatin. Obestatin is an endogenous ligand for the orphan G protein-coupled receptor GPR39 and is involved in satiety and decreased food intake. Also known as Appetite regulating hormone; GHRL; Growth hormone releasing peptide; Growth hormone secretagogue; M46 protein; Motilin related peptide; MTLRP; Obestatin; Obestatin preprohormone; PRO1066; UNQ524. Sequence notes: Gly-Ser-Ser-Phe-Leu-Ser-Pro- Glu-His-Gln-Lys-Ala-Gln-Gln-Arg-Lys-Glu- Ser-Lys-Lys-Pro-Pro-Ala-Lys-Leu-Gln-Pro- Arg (mo, rat).

Type: Primary
Antigen: Ghrelin 28
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 24-51/117
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR