Anti-Ghrelin 28 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-1375R-CY5
BOSSBS-1375R-CY5EA 883 EUR
BOSSBS-1375R-CY5
Anti-Ghrelin 28 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Ghrelin is an endogenous ligand for the growth hormone secretagogue receptor and is involved in regulating growth hormone release. Ghrelin is derived from a preprohormone called preproghrelin, which also generates a second peptide called obestatin. Obestatin is an endogenous ligand for the orphan G protein-coupled receptor GPR39 and is involved in satiety and decreased food intake. Also known as Appetite regulating hormone; GHRL; Growth hormone releasing peptide; Growth hormone secretagogue; M46 protein; Motilin related peptide; MTLRP; Obestatin; Obestatin preprohormone; PRO1066; UNQ524.More Product Information