Anti-H-1 beta Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss
BS-1407R-FITC
BOSSBS-1407R-FITCEA 900 EUR
BOSSBS-1407R-FITC
Anti-H-1 beta Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Required for activity of the Ah (dioxin) receptor. This protein is required for the ligand-binding subunit to translocate from the cytosol to the nucleus after ligand binding. The complex then initiates transcription of genes involved in the activation of PAH procarcinogens. The heterodimer with HIF1A or EPAS1/HIF2A functions as a transcriptional regulator of the adaptive response to hypoxia.

Type: Primary
Antigen: H-1 beta
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 681-789/789
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR