Anti-DEPDC1B Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-14278R-A750
BOSSBS-14278R-A750EA 874 EUR
BOSSBS-14278R-A750
Anti-DEPDC1B Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
GTPase activator activity.

Type: Primary
Antigen: DEPDC1B
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 451-529/529
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR