Anti-GNAI3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-1429R-A488
BOSSBS-1429R-A488EA 893 EUR
BOSSBS-1429R-A488
Anti-GNAI3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
GAI3 is the a subunit of inhibitory trimeric G protein (Gi), which inhibits adenylate cyclase once its G protein coupled receptors (GPCR) such as a2 adrenergic receptors are activated. GAI3 can be inactivated by pertussis toxin. Guanine nucleotide binding proteins are involved as modulators or transducers in various transmembrane signaling systems. G(k) is the stimulatory G protein of receptor regulated K(+) channels.

Type: Primary
Antigen: GNAI3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 488
Public Immunogen Range: 241-354/354
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR