Anti-DHDH Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-14302R-A680
BOSSBS-14302R-A680EA 890 EUR
BOSSBS-14302R-A680
Anti-DHDH Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
This gene encodes an enzyme that belongs to the family of dihydrodiol dehydrogenases, which exist in multiple forms in mammalian tissues and are involved in the metabolism of xenobiotics and sugars. These enzymes catalyze the NADP1-linked oxidation of transdihydrodiols of aromatic hydrocarbons to corresponding catechols. This enzyme is a dimeric dihydrodiol dehydrogenase, and it differs from monomeric dihydrodiol dehydrogenases in its high substrate specificity for trans-dihydrodiols of aromatic hydrocarbons in the oxidative direction. [provided by RefSeq, Jul 2008]

Type: Primary
Antigen: DHDH
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 281-334/334
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR