Anti-GRP10 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-14364R-A680
BOSSBS-14364R-A680EA 890 EUR
BOSSBS-14364R-A680
Anti-GRP10 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
GRP stimulates gastrin release as well as other gastrointestinal hormones. Operates as a negative feedback regulating fear and established a causal relationship between GRP-receptor gene expression, long-term potentiation, and amygdala-dependent memory for fear.
Type: Primary
Antigen: GRP10
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 24-52/146
Host: Rabbit
Isotype: IgG
Reactivity: Rat