Anti-GRP10 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-14364R-A750
BOSSBS-14364R-A750EA 890 EUR
BOSSBS-14364R-A750
Anti-GRP10 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
GRP stimulates gastrin release as well as other gastrointestinal hormones. Operates as a negative feedback regulating fear and established a causal relationship between GRP-receptor gene expression, long-term potentiation, and amygdala-dependent memory for fear.

Type: Primary
Antigen: GRP10
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 24-52/146
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR