Anti-MRP5/ABCC5 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-1437R-A488
BOSSBS-1437R-A488EA 893 EUR
BOSSBS-1437R-A488
Anti-MRP5/ABCC5 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
MRP5 (190-200 kDa) is closely related to MRP4, both lacking the first five membrane spanning regions. MRP5 is a GS-X multi specific organic anion pump (nucleotide analogs). MRP5 may transport DNP-GS and may be inhibited by certain inhibitors of organic anion transport (sulfinpyrazone). MRP5 may also transport organic anions with the anionic moiety of phosphate/phosphonate group, a function which provides the ability to resist against anti cancer drugs 6-MP and thioguanine as well as the anti-HIV drug PMEA.
Type: Primary
Antigen: MRP5/ABCC5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 488
Public Immunogen Range: 61-180/1436
Host: Rabbit
Isotype: IgG
Reactivity: Rat