Anti-HSP75/TRAP1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-1446R-A488
BOSSBS-1446R-A488EA 893 EUR
BOSSBS-1446R-A488
Anti-HSP75/TRAP1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Chaperone that expresses an ATPase activity. Involved in maintaining mitochondrial function and polarization, most likely through stabilisation of mitochondrial complex I. Is a negative regulator of mitochondrial respiration able to modulate the balance between oxidative phosphorylation and aerobic glycolysis. The impact of TRAP1 on mitochondrial respiration is probably mediated by modulation of mitochondrial SRC and inhibition of SDHA.

Type: Primary
Antigen: HSP75/TRAP1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 488
Public Immunogen Range: 501-650/704
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR