Anti-Prohibitin Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-1449R-CY3
BOSSBS-1449R-CY3EA 909 EUR
BOSSBS-1449R-CY3
Anti-Prohibitin Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Prohibitin inhibits DNA synthesis. It has a role in regulating proliferation. As yet it is unclear if the protein or the mRNA exhibits this effect. May play a role in regulating mitochondrial respiration activity and in aging.

Type: Primary
Antigen: Prohibitin
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range: 171-272/272
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Others
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR