Anti-MTHFR Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-1484R-A555
BOSSBS-1484R-A555EA 893 EUR
BOSSBS-1484R-A555
Anti-MTHFR Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
catalyses the conversion of 5,10-methylenetetrahydrofolate to 5-methyltetrahydrofolate, a co-substrate for homocysteine remethylation to methionine.
Type: Primary
Antigen: MTHFR
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 555
Public Immunogen Range: 165-300/656
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen