Anti-MTHFR Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-1484R-A680
BOSSBS-1484R-A680EA 890 EUR
BOSSBS-1484R-A680
Anti-MTHFR Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
catalyses the conversion of 5,10-methylenetetrahydrofolate to 5-methyltetrahydrofolate, a co-substrate for homocysteine remethylation to methionine.

Type: Primary
Antigen: MTHFR
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 165-300/656
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR