Anti-AQP1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-1506R-CY3
BOSSBS-1506R-CY3EA 909 EUR
BOSSBS-1506R-CY3
Anti-AQP1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Forms a water-specific channel that provides the plasma membranes of red cells and kidney proximal tubules with high permeability to water, thereby permitting water to move in the direction of an osmotic gradient.

Type: Primary
Antigen: AQP1
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range: 181-269/269
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR