Anti-C20orf134 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-15093R-A488
BOSSBS-15093R-A488EA 893 EUR
BOSSBS-15093R-A488
Anti-C20orf134 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
C20orf134 is a 245 amino acid secreted protein that belongs to the Actin family and is encoded by a gene which maps to human chromosome 20. Comprising approximately 2% of the human genome, chromosome 20 contains nearly 63 million bases that encode over 600 genes, some of which are associated with Creutzfeldt-Jakob disease, amyotrophic lateral sclerosis, spinal muscular atrophy, ring chromosome 20 epilepsy syndrome and Alagille syndrome. Additionally, chromosome 20 contains a region with numerous genes which are thought important for seminal production and may be potential targets for male contraception.

Type: Primary
Antigen: C20orf134
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 488
Public Immunogen Range: 21-100/245
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR