Anti-BMPR1A/CD292 Rabbit Polyclonal Antibody (HRP (Horseradish PE (Phycoerythrin)rOxidase))

Lieferant: Bioss
BS-1509R-HRP
BOSSBS-1509R-HRPEA 900 EUR
BOSSBS-1509R-HRP
Anti-BMPR1A/CD292 Rabbit Polyclonal Antibody (HRP (Horseradish PE (Phycoerythrin)rOxidase))
Antikörper
On ligand binding, forms a receptor complex consisting of two type II and two type I transmembrane serine/threonine kinases. Type II receptors phosphorylate and activate type I receptors which autophosphorylate, then bind and activate SMAD transcriptional regulators. Receptor for BMP-2 and BMP-4.

Type: Primary
Antigen: BMPR1A/CD292
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range:101-200/532
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR