Anti-C2GNT3 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-15145R
BOSSBS-15145REA 725 EUR
BOSSBS-15145R
Anti-C2GNT3 Rabbit Polyclonal Antibody
Antikörper
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: C2GNT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR