Anti-Pan-ras Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-1515R
BOSSBS-1515REA 725 EUR
BOSSBS-1515R
Anti-Pan-ras Rabbit Polyclonal Antibody
Antikörper
RAS superfamily comprises around 50 related genes encoding GTP-binding domain (G-proteins) involved in signal transduction. The main genes are HRAS, NRAS and KRAS. Ras proteins are membrane-bound GTPases. The inactive form is GDP-bound. They are activated by ligand-binding receptor tyrosine kinases such as EGFR, PDGFR, colony-stimulating factor and fibroblast growth factor. These kinases transiently convert RAS-GDP to RAS-GTP, the active form of RAS. Single amino acid substitutions can activate RAS making it highly oncogenic. Such mutations generally reduce the GTPase activity of RAS, prolonging it in its active GTP-bound form. The consequence of this is sustained activation of the RAF1-MAPK signalling pathway. RAS mutations are found in 10-15% of tumours. A high incidence of RAS mutations is found in pancreatic cancers.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Pan-ras
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR