Anti-C2orf76 Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-15160R
BOSSBS-15160REA 725 EUR
BOSSBS-15160R
Anti-C2orf76 Rabbit Polyclonal Antibody
Antikörper
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: C2orf76
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit