Anti-C4orf17 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-15187R-CY7
BOSSBS-15187R-CY7EA 893 EUR
BOSSBS-15187R-CY7
Anti-C4orf17 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
C4orf17 (chromosome 4 open reading frame 17) is a 359 amino acid protein encoded by a gene that maps to human chromosome 4. Human chromosome 4 represents approximately 6% of the human genome and contains nearly 900 genes. Notably, the Huntingtin gene, which is found to encode an expanded glutamine tract in cases of Huntington's disease, is on chromosome 4. FGFR-3 is also encoded by a gene that is located on chromosome 4, and has been associated with thanatophoric dwarfism, achondroplasia, Muenke syndrome and bladder cancer. Chromosome 4 is also tied to Ellis-van Creveld syndrome, methylmalonic acidemia and polycystic kidney disease.

Type: Primary
Antigen: C4orf17
Clonality: Polyclonal
Clone:
Conjugation: Cy7
Public Immunogen Range: 121-220/359
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR