Anti-C4orf29 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-15190R-A350
BOSSBS-15190R-A350EA 893 EUR
BOSSBS-15190R-A350
Anti-C4orf29 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
C4orf29 (chromosome 4 open reading frame 29) is a 414 amino acid secreted protein that exists as four alternatively spliced isoforms and is encoded by a gene that maps to human chromosome 4. Human chromosome 4 represents approximately 6% of the human genome and contains nearly 900 genes. Notably, the Huntingtin gene, which is found to encode an expanded glutamine tract in cases of Huntington's disease, is located on chromosome 4. FGFR-3 is also encoded by a gene that maps to human chromosome 4 and has been associated with thanatophoric dwarfism, achondroplasia, Muenke syndrome and bladder cancer. Chromosome 4 is also tied to Ellis-van Creveld syndrome, methylmalonic acidemia and polycystic kidney disease.

Type: Primary
Antigen: C4orf29
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 350
Public Immunogen Range: 121-220/414
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR