Anti-C4orf29 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Lieferant: Bioss
BS-15190R-FITC
BOSSBS-15190R-FITCEA 900 EUR
BOSSBS-15190R-FITC
Anti-C4orf29 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
C4orf29 (chromosome 4 open reading frame 29) is a 414 amino acid secreted protein that exists as four alternatively spliced isoforms and is encoded by a gene that maps to human chromosome 4. Human chromosome 4 represents approximately 6% of the human genome and contains nearly 900 genes. Notably, the Huntingtin gene, which is found to encode an expanded glutamine tract in cases of Huntington's disease, is located on chromosome 4. FGFR-3 is also encoded by a gene that maps to human chromosome 4 and has been associated with thanatophoric dwarfism, achondroplasia, Muenke syndrome and bladderMore Product Information