Anti-C4orf46 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-15194R-CY3
BOSSBS-15194R-CY3EA 893 EUR
BOSSBS-15194R-CY3
Anti-C4orf46 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Representing approximately 6% of the human genome, chromosome 4 contains nearly 900 genes. Notably, the Huntingtin gene, which is found to encode an expanded glutamine tract in cases of Huntington's disease, is on chromosome 4. FGFR-3 is also encoded on chromosome 4 and has been associated with thanatophoric dwarfism, achondroplasia, Muenke syndrome and bladder cancer. Chromosome 4 is also tied to Ellis-van Creveld syndrome, methylmalonic acidemia and polycystic kidney disease. Chromosome 4 reportedly contains the largest gene deserts (regions of the genome with no protein encoding genes) and has one of the two lowest recombination frequencies of the human chromosomes. The C4orf46 gene product has been provisionally designated C4orf46 pending further characterization.

Type: Primary
Antigen: C4orf46
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range: 51-113/113
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR