Anti-CD7 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-1520R-A680
BOSSBS-1520R-A680EA 890 EUR
BOSSBS-1520R-A680
Anti-CD7 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
CD7 is a 40 kDa transmembrane, single-chain glycoprotein, which is a member of the immunoglobulin superfamily. CD7 is expressed by the majority of thymocytes and mature T cells, NK cells and pre-B cells. It plays an essential role in T-cell interactions and also in T-cell/B-cell interaction during early lymphoid development. The function of CD7 is not yet known although cross-linking of CD7 with induces a T cell transmembrane calcium flux and CD7 expression is induced by ionomycin. The CD7 molecule has been reported to be the receptor for the IgM-Fc portion (FcR mu) on the surface of T cells.

Type: Primary
Antigen: CD7
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 181-240/240
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR