Anti-C6orf106 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-15213R-CY5
BOSSBS-15213R-CY5EA 900 EUR
BOSSBS-15213R-CY5
Anti-C6orf106 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
C6orf106 is a Making up nearly 6% of the human genome, chromosome 6 contains around 1,200 genes within 170 million base pairs of sequence. Deletion of a portion of the q arm of chromosome 6 is associated with early onset intestinal cancer suggesting the presence of a cancer susceptibility locus. Porphyria cutanea tarda is associated with chromosome 6 through the HFE gene which, when mutated, predisposes an individual to developing this porphyria. Notably, the PARK2 gene, which is associated with Parkinson's disease, and the genes encoding the major histocompatiblity complex proteins, which are key molecular components of the immune system and determine predisposition to rheumatic diseases, are also located on chromosome 6. Stickler syndrome, 21-hydroxylase deficiency and maple syrup urine disease are also associated with genes on chromosome 6. A bipolar disorder susceptibility locus has been identified on the q arm of chromosome 6. The C6orf106 gene product has been provisionally designated C6orf106 pending further characterization.

Type: Primary
Antigen: C6orf106
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 1-80/298
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR