Anti-C6ORF154 Rabbit Polyclonal Antibody (FITC)

Lieferant: Bioss
BS-15226R-FITC
BOSSBS-15226R-FITCEA 900 EUR
BOSSBS-15226R-FITC
Anti-C6ORF154 Rabbit Polyclonal Antibody (FITC)
Antikörper
Making up nearly 6% of the human genome, chromosome 6 contains around 1200 genes within 170 million base pairs of sequence. Deletion of a portion of the q arm of chromosome 6 is associated with early onset intestinal cancer suggesting the presence of a cancer susceptibility locus. Porphyria cutanea tarda is associated with chromosome 6 through the HFE gene which, when mutated, predisposes an individual to developing this porphyria. Notably, the PARK2 gene, which is associated with Parkinson's disease, and the genes encoding the major histocompatiblity complex proteins, which are key molecular components of the immune system and determine predisposition to rheumatic diseases, are also located on chromosome 6. Stickler syndrome, 21-hydroxylase deficiency and maple syrup urine disease are also associated with genes on chromosome 6. A bipolar disorder susceptibility locus has been identified on the q arm of chromosome 6. The C6orf154 gene product has been provisionally designated C6orf154 pending further characterisation.

Type: Primary
Antigen: C6ORF154
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 51-150/316
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR