Anti-mucin3/Muc3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-1525R-A680
BOSSBS-1525R-A680EA 899 EUR
BOSSBS-1525R-A680
Anti-mucin3/Muc3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Major glycoprotein component of a variety of mucus gels. Thought to provide a protective, lubricating barrier against particles and infectious agents at mucosal surfaces. May be involved in ligand binding and intracellular signaling.

Type: Primary
Antigen: mucin3/Muc3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1501-1600/1719
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR