Anti-mucin3/Muc3 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-1525R-CY5
BOSSBS-1525R-CY5EA 909 EUR
BOSSBS-1525R-CY5
Anti-mucin3/Muc3 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Major glycoprotein component of a variety of mucus gels. Thought to provide a protective, lubricating barrier against particles and infectious agents at mucosal surfaces. May be involved in ligand binding and intracellular signaling.
Type: Primary
Antigen: mucin3/Muc3
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 1501-1600/1719
Host: Rabbit
Isotype: IgG
Reactivity: Human