Anti-HBQ1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 350)

Lieferant: Bioss
BS-15419R-A350
BOSSBS-15419R-A350EA 893 EUR
BOSSBS-15419R-A350
Anti-HBQ1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 350)
Antikörper
Theta-globin mRNA is found in human fetal erythroid tissue but not in adult erythroid or other nonerythroid tissue. The theta-1 gene may be expressed very early in embryonic life, perhaps sometime before 5 weeks. Theta-1 is a member of the human alpha-globin gene cluster that involves five functional genes and two pseudogenes. The order of genes is: 5' - seta - pseudoseta - mu - pseudoalpha-2 -pseudoalpha-1 - alpha-2 - alpha-1 - theta-1 - 3'. Research supports a transcriptionally active role for the gene and a functional role for the peptide in specific cells, possibly those of early erythroid tissue.

Type: Primary
Antigen: HBQ1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 350
Public Immunogen Range: 1-100/142
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR